missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle
GREENER_CHOICE

Product Code. 10186700
Change view
Click to view available options
Nominal Capacity:
175 mL
Max. Force:
27,500 x g
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Nominal Capacity Max. Force unitSize
10186700 175 mL 27,500 x g Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10186700 Supplier Thermo Scientific™ Supplier No. 31440175
  • ONLINE EXCLUSIVE PRICE
    117.55 €
    111.77 € / Pack of 4
    Save 5.78 €
    4% Off
View all promotions
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Process pelleting applications such as cell culture harvesting and protein biochemistry with this polycarbonate conical-bottom centrifuge bottle.

Process pelleting applications such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations, with Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle. Molded of rigid and robust polycarbonate copolymer, these transparent bottles are autoclavable and offer excellent mechanical strength.

Features:
  • For low to moderate speed centrifugation of biological and chemical samples
  • Includes polypropylene sealing cap assembly with silicone gasket for added leakproof† assurance at maximum speeds or when spinning hazardous materials
  • Must be used with Nalgene adapter for conical-bottom centrifuge bottles (DS3126-0175)
  • For proper performance, bottles must be filled to 80% of total capacity
  • Autoclavable, transparent, leakproof† when used with supplied Nalgene closure, molded-in graduations
  • Autoclaving will shorten the life of PC centrifuge bottles
Note: Not recommended to use in Thermo Fisher Fiberlite F14-6x250y rotor, due to the cavity depth.

You can achieve successful leakproof† performance using these two bottles in Thermo Fisher Fiberlite F12-6x500 LEX rotor when used with the 010-0152 adaptor at speeds up to but not exceeding 24,470 x g.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 175 mL
Color Clear
Material Polycarbonate
No. per Case 36
No. per Pack 4
Nominal Capacity 175 mL
Graduated Yes
Max. Force 27,500 x g
Height (Metric) 140.5 mm
Closure Material Polypropylene, red rubber
Diameter (Metric) 61.8 mm
Closure Type Polypropylene Sealing Closure
Through the Closure Dispensing No
Autoclavable Autoclavable
Capacity (English) 5.91 oz.
Shape Conical
Product Type Centrifuge Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.