missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate, End Cap

Product Code. 10090282
Change view
Click to view available options
Quantity:
Case of 4
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10090282 Case of 4 Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10090282 Supplier Thermo Scientific™ Supplier No. 26882075
  • ONLINE EXCLUSIVE PRICE
    36.80 €
    34.70 € / Pack of 4
    Save 2.10 €
    5% Off
View all promotions
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use With: Nalgene Carboy 2630 and 2261

Sanitary ports for easy fill/ing and dispensing. Thermo Scientific™ Nalgene™ End Caps in polycarbonate or polypropylene to provide operations. Caps also can be securely mounted to 3 in. sanitary flange for applications used with Nalgene™ Sanitary Carboys.

  • Autoclavable
  • Compatible with Nalgene Sanitary Carboys (Cat. Nos. 2630 and 2261)
TRUSTED_SUSTAINABILITY

Specifications

Description Two 0.75 in. (19mm) sanitary ports, polycarbonate
Diameter (English) Inner Tubing 0.75 in.
Diameter (Metric) Inner Tubing 19 mm
For Use With (Equipment) Nalgene Carboy 2630 and 2261
Material Polycarbonate
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type End Cap
Quantity Case of 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.