missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels
GREENER_CHOICE

Product Code. 11333482
Change view
Click to view available options
Height (Metric):
13 mm
20 mm
22 mm
25 mm
51 mm
Width (Metric):
22 mm
38 mm
44 mm
67 mm
102 mm
Quantity:
1500 Cs.
3250 Cs.
4000 Cs.
500 Cs.
Unit Size:
Pack of 300
Pack of 3250
Pack of 4000
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Quantity unitSize
11333482 22 mm 22 mm 4000 Cs. Pack of 4000
10713052 20 mm 38 mm 3250 Cs. Pack of 3250
11319821 25 mm 67 mm 1500 Cs. Pack of 300
11394555 13 mm 44 mm 4000 Cs. Pack of 4000
11353482 51 mm 102 mm 500 Cs. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11333482 Supplier Thermo Scientific™ Supplier No. 63190010
Stock Up Your Lab
Use promo code 28007
View all promotions
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.

Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.

Features:

  • Durable polyolefin labels
  • Rated to −29°C
  • Resistant to ultraviolet light, mildew, chemicals and dirt, and won't discolor, crack or tear
  • Excellent for labeling microscopic slides—adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents, although solvents may dissolve adhesive
  • Autoclavable
  • Formatted for easy setup in word processing programs; instructions included in each pack

Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.

TRUSTED_SUSTAINABILITY

Specifications

Description Polyolefin Label, 0.9 x 0.9
Material Polyolefin
Product Type Label
Labels per Sheet 80
No. per Case 4000/Cs.
No. per Pack 800/Pk.
Quantity 4000 Cs.
Width (English) 0.9 in.
Width (Metric) 22 mm
Height (English) 0.9 in.
Height (Metric) 22 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.