missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
GREENER_CHOICE

Product Code. 10010741
Change view
Click to view available options
Diameter (Metric)Top:
35 mm
54 mm
63 mm
76 mm
98 mm
158 mm
Capacity (Metric):
10 mL
41 mL
64 mL
114 mL
254 mL
976 mL
Unit Size:
Pack of 12
Pack of 2
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Capacity (Metric) Unit Size
10010741 35 mm 10 mL Pack of 12
10562131 54 mm 41 mL Pack of 12
10490143 63 mm 64 mL Pack of 12
10294491 76 mm 114 mL Pack of 6
10284541 98 mm 254 mL Pack of 4
10505981 158 mm 976 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10010741 Supplier Thermo Scientific™ Supplier No. 42500035
  • ONLINE EXCLUSIVE PRICE
    39.95 €
    35.00 € / Pack of 12
    Save 4.95 €
    12% Off
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric)Top 35 mm
Capacity (Metric) 10 mL
Diameter (Metric) Stem 6 mm
Material Polypropylene
Autoclavable Autoclavable
Description Funnel Analytical PP 35mm
Length (Metric) Stem 50 mm
Quantity 12/Pk.; 36/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.