missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels

Product Code. 10284541
Change view
Click to view available options
Diameter (Metric)Top:
35 mm
54 mm
63 mm
76 mm
98 mm
158 mm
Capacity (Metric):
10 mL
41 mL
64 mL
114 mL
254 mL
976 mL
Unit Size:
Pack of 12
Pack of 2
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Capacity (Metric) Unit Size
10284541 98 mm 254 mL Pack of 4
10010741 35 mm 10 mL Pack of 12
10562131 54 mm 41 mL Pack of 12
10490143 63 mm 64 mL Pack of 12
10294491 76 mm 114 mL Pack of 6
10505981 158 mm 976 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10284541 Supplier Thermo Scientific™ Supplier No. 42500100
  • ONLINE EXCLUSIVE PRICE
    31.60 €
    30.84 € / Pack of 4
    Save 0.76 €
    2% Off
Visualizza tutte le promozioni
In Stock
Aggiungi al carrello
Edge
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifica

Diameter (Metric)Top 98 mm
Capacity (Metric) 254 mL
Diameter (Metric) Stem 9 mm
Material Polypropylene
Autoclavable Autoclavable
Description Funnel Analytical PP 100mm
Length (Metric) Stem 100 mm
Quantity 4/Pk.; 24/Cs.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.