missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
GREENER_CHOICE

Product Code. 10587951
Change view
Click to view available options
Nominal Capacity:
250 mL
450 mL
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Nominal Capacity Unit Size
10587951 450 mL Pack of 4
10430613 250 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587951 Supplier Thermo Scientific™ Supplier No. 31410500
  • ONLINE EXCLUSIVE PRICE
    146.75 €
    135.17 € / Pack of 4
    Save 11.58 €
    7% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with these PPCO centrifuge bottles, which come with a sealing closure.

Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles, which include a sealing closure for added leakproof† assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Additionally, these PPCO bottles are autoclavable and offer excellent mechanical strength.

Features:
  • For low- to moderate-speed centrifugation of biological and chemical samples
  • Bottle includes polypropylene sealing cap assembly with silicone gasket for added leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity

Requires:

When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 450 mL
Color Translucent
Material PPCO
Length (Metric) 160 mm
No. per Pack 4
Nominal Capacity 450 mL
Closure Included Yes
Graduated No
Max. Force 13,700 x g
Height (Metric) 159.3 mm
Width (English) 2.735 in.
Length (English) 2.735 in.
Height (English) 6.27 in.
Closure Material Polypropylene, red rubber
Diameter (Metric) 69.5 mm
Closure Type Sealing Cap Assembly with Silicone Gasket
Through the Closure Dispensing No
Autoclavable Autoclavable
Capacity (English) 16.9 oz.
Shape Round
Product Type Centrifuge Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.