missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
GREENER_CHOICE

Product Code. 10374061
Change view
Click to view available options
Capacity (Metric):
15 mL
50 mL
Length (Metric):
118.6 mm
133.1 mm
Diameter (Metric):
16.8 mm
28.7 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Diameter (Metric) unitSize
10374061 15 mL 118.6 mm 16.8 mm Pack of 10
10602682 50 mL 133.1 mm 28.7 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374061 Supplier Thermo Scientific™ Supplier No. 31030015
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Simplify measurements using these PPCO conical-bottom centrifuge tubes, which feature molded-in graduations. Translucent PPCO has excellent chemical resistance.

Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.

Features:

  • Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
  • Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (tubes must be filled at least 80% for proper performance)
  • Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
  • Molded-in graduations last the life of the tube
  • Last longer than polycarbonate tubes under conditions of repeated autoclaving
  • Tube are both autoclavable and graduated

Ordering Information:

Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 15 mL
Material Polypropylene Copolymer
No. per Case 100
No. per Pack 10
Sterility Non-sterile
Autoclavable Autoclavable
Shape Conical
Length (Metric) 118.6 mm
Bottom Shape Conical
Writing Area No
Diameter (Metric) 16.8 mm
Graduations Molded-in
Product Type Centrifuge Tube
Max. RCF 6000 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.