Learn More
Thermo Scientific™ Nalgene™ PVDF True Union Clamps
Description
Repeatedly and reliably seal carboys with Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gaskets and end caps. Molded of PVDF fluoride, these threaded clamps are a fully autoclavable clamp system.
- Available in three sizes and with Mini and Tri-Clover clamp types
- Threaded and autoclavable
- Recommended for use with Nalgene™ Sanitary Carboys (Cat. Nos. 2261, 2630 and 2640) and Nalgene™ End Caps (Cat. Nos. 2688 and 2689)
- Recommended for use with Nalgene™ Autoclavable Carboys with Spigot (Cat. No. 2319)
Specifications
Specifications
| Quantity | 1/Pk., 1/Cs. |
| Type | Tri Clover Tube Clamp |
| Material | PVDF |
| For Use With (Equipment) | Nalgene™ Sanitary Carboys, Nalgene™ Autoclavable Carboys with Spigot |
| Size (English) Grip | 1.5 in. |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.