missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
GREENER_CHOICE

Product Code. p-9078039
Click to view available options
Capacity (Metric):
9 L
20 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10633521

Brand: Thermo Scientific™ 23220020

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

For Use With: Ideal for use when visual inspection of contents is necessary; Autoclavable; Closure Material: Polypropylene; Color: Clear

Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot, which features a wide-mouth opening, a sturdy stainless-steel handle and space-saving rectangular design. Clear construction allows easy viewing of carboy contents. Super tough; impact resistant. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features

  • Molded-in graduations in 1 gallon and 5L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Molded-in spigot boss provides strength and leak resistance
  • A clear wall makes this carboy ideal when visual inspection of contents is necessary
  • Follow autoclave instructions provided with the product for proper function; tubing adapter is not autoclavable
  • Autoclavable/Transparent/Graduated

Includes:

Polypropylene spigot and screw closure

TRUSTED_SUSTAINABILITY

Specifikationer

Autoclavable Autoclavable
Capacity (English) 2.4 gal.
Capacity (Metric) 9 L
Closure Material Polypropylene
Closure Size 100 mm
Color Transparent
Description Autoclavable Clear Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck 86 mm
For Use With (Application) Ideal for use when visual inspection of contents is necessary
Height (Metric) With Closure 361 mm
Includes Molded-in spigot boss w/tubing adapter
Length (Metric) 220 mm
Material Polycarbonate
No. per Case 6/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Spigot
Width (Metric) 153 mm
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.