missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Replacement Silicone O-Ring for Centrifuge Bottle Sealing Cap
GREENER_CHOICE

Product Code. 11795223
Change view
Click to view available options
Product Type:
O-ring
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
11795223 O-ring Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11795223 Supplier Thermo Scientific™ Supplier No. 713141PPCO
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use Nalgene™ Silicone O-Ring as a replacement part in centrifuge bottle sealing caps.

The Nalgene™ Replacement Silicone O-Ring is used as a replacement for the sealing cap on Nalgene™ Cat #3141-1006 Super-Speed Centrifuge Bottle.

  • Replacement O-ring
  • Used for cap of Nalgene™ Super-Speed Centrifuge Bottle #3141-1006
  • Silicone
  • 85mm size
  • Red
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Silicone
For Use With (Equipment) Nalgene Cat #3141-1006 Super-Speed Centrifuge Bottle Cap
Closure Type Sealing Cap
Shape Round
Product Type O-ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.