missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile

Product Code. 10156192
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
2800 mL
Closure Size:
70 mm
38-430 mm
45-430 mm
Closure Type:
Solid, non-vented
Vented
Unit Size:
Pack of 12
Pack of 24
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 10156192

Brand: Thermo Scientific™ 41152800

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.

Save preparation time and avoid contamination risk with Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features

  • Sterile and single use for reduced contamination risk
  • BPA-free
  • Collapses when autoclaved, reducing biohazardous waste
  • Graduated for quick volume assessment
  • Regulatory documentation available on request

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 95 oz.
Capacity (Metric) 2800 mL
Material PETG
No. per Case 4
No. per Pack 1
Shape Erlenmeyer
Surface Coating None
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Mouth Style Wide
Sterility Sterile
Bottom Shape Plain
Closure Type Vented
Closure Size 70 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.