missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile

Product Code. 10266432
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
2800 mL
Closure Size:
70 mm
38-430 mm
45-430 mm
Closure Type:
Solid, non-vented
Vented
Unit Size:
Pack of 12
Pack of 24
Pack of 4
Pack of 6
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Closure Type Unit Size
10266432 125 mL 38-430 mm Vented Pack of 24
10592033 250 mL 38-430 mm Vented Pack of 12
10686673 500 mL 45-430 mm Vented Pack of 12
10357302 1000 mL 45-430 mm Vented Pack of 6
10473582 2000 mL 45-430 mm Vented Pack of 4
10156192 2800 mL 70 mm Vented Pack of 4
10465622 125 mL 38-430 mm Solid, non-vented Pack of 24
10292032 250 mL 38-430 mm Solid, non-vented Pack of 12
10179661 500 mL 45-430 mm Solid, non-vented Pack of 12
10475622 1000 mL 45-430 mm Solid, non-vented Pack of 6
10385012 2000 mL 45-430 mm Solid, non-vented Pack of 4
10228732 2800 mL 70 mm Solid, non-vented Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10266432 Supplier Thermo Scientific™ Supplier No. 41150125
  • ONLINE EXCLUSIVE PRICE
    380.28 €
    247.00 € / Pack of 24
    Save 133.28 €
    35% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.

Save preparation time and avoid contamination risk with Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features

  • Sterile and single use for reduced contamination risk
  • BPA-free
  • Collapses when autoclaved, reducing biohazardous waste
  • Graduated for quick volume assessment
  • Regulatory documentation available on request

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 4 oz.
Capacity (Metric) 125 mL
Material PETG
No. per Case 24
No. per Pack 1
Shape Erlenmeyer
Surface Coating None
Green Features ACT Ecolabel (Silver)
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Mouth Style Narrow
Sterility Sterile
Bottom Shape Plain
Closure Type Vented
Closure Size 38-430 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.