missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Spigots for Storage Tanks

Product Code. 12867830
Change view
Click to view available options
Material:
LDPE
Polypropylene
Closure Size:
1.12 in. x 12 female screw thread
12.7mm NPT male thread
19mm NPT male screw thread
Unit Size:
Pack of 12
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Closure Size unitSize
12867830 LDPE 12.7mm NPT male thread Pack of 6
12877830 LDPE 19mm NPT male screw thread Pack of 6
15981651 Polypropylene 1.12 in. x 12 female screw thread Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12867830 Supplier Thermo Scientific™ Supplier No. 64200500
  • ONLINE EXCLUSIVE PRICE
    297.00 €
    279.00 € / Pack of 6
    Save 18.00 €
    6% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ensure safe and efficient liquid dispensing with Thermo Scientific™ Nalgene™ Spigots for Storage Tanks, for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.

Ensure safe and efficient liquid dispensing from tanks by using Thermo Scientific™ Nalgene™ Spigots for Storage Tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.

Includes

  • PTFE O-rings (6421-0010 includes two O-rings)

Note:
Make sure container and spigot have compatible chemical resistance before installing.

TRUSTED_SUSTAINABILITY

Specifications

Closure Size 12.7mm NPT male thread
Type Spigot
Material LDPE
Capacity (English) 100 gal.
Capacity (Metric) 378 L
No. per Case 6
No. per Pack 1

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.