missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure

Product Code. 10567112
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20 mm
24 mm
38-430
Unit Size:
Pack of 12
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Unit Size
10567112 60 mL 24 mm Pack of 12
10451181 30 mL 20 mm Pack of 12
10004710 125 mL 38-430 Pack of 12
10064740 250 mL 38-430 Pack of 12
10637711 500 mL 38-430 Pack of 12
10647711 1 L 38-430 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10567112 Supplier Thermo Scientific™ Supplier No. 20180060
  • ONLINE EXCLUSIVE PRICE
    22.65 €
    20.50 € / Pack of 12
    Save 2.15 €
    9% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.

Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Square sides allow for good use of space and an easy grip
  • Caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof† when used with Nalgene closures
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Leakproof† Polypropylene Screw Caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Recycling Information:

Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Color Translucent
Description Narrow-Mouth Square Bottle
Diameter (English) 1.6 in.
Diameter (English) Inner Neck 0.7 in.
Diameter (Metric) Inner Neck 18 mm
Includes Leakproof polypropylene screw closures
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural, translucent
Graduations Yes
Closure Included Yes
Graduated Yes
Mouth Narrow
ACT Label Yes
Height (Metric) 83 mm
Width (English) 1.61 in.
Height (English) 3.2 in.
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 41 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Not Autoclavable
Capacity (English) 2 oz.
Shape Square
Certifications/Compliance USP Class VI, FDA - 21 CFR
Product Type Square Narrow Mouth Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.