missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
BIOPHARMA

Product Code. 10384333
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
53B
Unit Size:
Pack of 12
Pack of 200
Pack of 24
Pack of 280
Pack of 40
Pack of 60
Pack of 96
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Unit Size
10384333 125 mL 38-430 mm Pack of 96
10561075 30 mL 20-415 mm Pack of 280
10304293 60 mL 24-415 mm Pack of 200
10140463 250 mL 38-430 mm Pack of 60
10262953 500 mL 38-430 mm Pack of 40
10622634 1 L 38-430 mm Pack of 24
10222813 2 L 53B Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10384333 Supplier Thermo Scientific™ Supplier No. 3220200125
  • ONLINE EXCLUSIVE PRICE
    247.00 €
    227.00 € / Pack of 96
    Save 20.00 €
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.

Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.

  • Heavy-walled and durable
  • Shatter-resistant
  • Provide excellent gas barrier properties
  • In shrink-wrap trays
  • Molded in graduations
  • Clear bottles for easy identification of contents
  • Nonsterile
  • Leakproof†

Note:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Color Clear
Description 125mL, 38-430 closure, tray pack
Diameter (English) 2.12 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Storing Media and Sera
Material PETG
No. per Case 96
No. per Pack 24
Sterility Non-sterile
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 110 mm
Height (English) 4.3 in.
Closure Material HDPE
Closure Size 38-430 mm
Diameter (Metric) 54 mm
Capacity (English) 4.2 oz.
Shape Square
Certifications/Compliance USP Class VI, FDA Compliance 21 CFR
Product Type Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.