missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure

Product Code. p-4521806
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
1 L
2000 mL
Unit Size:
Pack of 12
Pack of 24
Pack of 48
Pack of 96

Product Code. 10037950

Brand: Thermo Scientific™ 20190030

Only null left
Add to basket
Add to basket

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.

Features:

  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Suitable for frozen storage down to -40°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Sterile and non-pyrogenic
  • Available in low particulate format
  • Supported by extensive validation documentation

Ordering Information:

  • Packed in shrink-wrapped trays

Note:

  • 2 L size has molded-in handgrips

Warranty:

  • 90 days
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.496 in.
Diameter (English) Inner Neck 0.55 in.
Diameter (Metric) Inner Neck 14 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage
Material PETG
No. per Case 96/Cs.
No. per Pack 24/Pk.
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Certifications/Compliance Bottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines.
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 64 mm
Product Type Media Bottle
Height (English) 2.52 in.
Closure Material HDPE
Closure Size 20 mm
Diameter (Metric) 38 mm
Autoclavable Not Autoclavable
Workflow Step Upstream media/buffer prep and storage. Upstream process solution storage. Upstream Validation
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.