missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack

Product Code. 30117594
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
Unit Size:
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck Finish Unit Size
30117594 4 mL 13-415 mm Pack of 2000
10100713 8 mL 20-415 mm Pack of 2000
10610144 15 mL 20-415 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30117594 Supplier Thermo Scientific™ Supplier No. 3120849125
Estimated Shipment: 11-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect light-sensitive samples with translucent amber HDPE diagnostic bottles, intended for storage and shipping of small-volume, light-sensitive, valuable reagents.

Protect light-sensitive samples with Thermo Scientific™ Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure, suitable for storage and shipping of small-volume, light-sensitive, valuable reagents. Translucent bottles provide light resistance with the ability to see liquid level and are ideal for test kits and many small-volume applications.

Features:
  • Nonsterile, high-density polyethylene bottles
  • Opaque, linerless polypropylene closures
  • Durable, uniform walls that resist splitting and puncturing
  • Non-cytotoxic
  • Leakproof†

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton; 2,000 per case.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.125 oz., 4 mL
Capacity (Metric) 4 mL
Color Translucent Amber
Description 3.4mL, 20-415 closure, bulk pack
Diameter (Metric) Inner Neck 8 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 13-415 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 41 mm
Width (English) 0.62 in.
Height (English) 1.58 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 13-415 mm
Diameter (Metric) 16 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Capacity (English) 0.125 oz.
Shape Round
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Product Type Sample Container
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.