missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid, 99%

Product Code. 11436117
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436117 5 g 5g
11446117 25 g 25g
11456117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11436117 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B25096.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 389-08-2 | C12H12N2O3 | 232.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Nalidixic acid is a synthetic 1,8-naphthyridine antimicrobial agent. It is also used as an inhibitor of bacterial DNA gyrase. It plays an important role in the treatment of urinary tract infections caused by susceptible gram-negative microorganisms, including the majority of E. Coli, Enterobacter species, Klebsiella species and Proteus species.

Solubility
Soluble in diethyl ether, acetone, methylene chloride, chloroform and methanol. Insoluble in water.

Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389-08-2
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.239
MDL Number MFCD00006884
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
PubChem CID 4421
ChEBI CHEBI:100147
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O

Specifications

Melting Point 228°C to 230°C
Quantity 5 g
Beilstein 750515
Merck Index 14,6359
Solubility Information Soluble in diethyl ether,acetone,methylene chloride,chloroform and methanol. Insoluble in water.
Formula Weight 232.24
Percent Purity 99%
Chemical Name or Material Nalidixic acid
compliance-icons compliance-icons
Product Identifier
  • Nalidixic acid
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Nalidixic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.