missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid sodium salt

Product Code. 15455749
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455749 25 g 25g
15445749 5 g 5g
15465749 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15455749 Supplier Thermo Scientific Alfa Aesar Supplier No. J63550.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Nalixidic acid, CAS # 389-08-2, is a quinolone antibiotic that inhibits cell division. | CAS: 3374-05-8 | C12H11N2NaO3 | 254.22 g/mol

Nalidixic acid sodium salt is an antibacterial agent, which is inhibitor of bacterial DNA polymerase & avian myeloblastoma virus reverse transcriptase. The compound inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nalidixic acid is a monocarboxylic acid and quinoline antibiotic
• Nalixidic acid is a DNA synthesis inhibitor that acts by preventing DNA gyrase activity

Application

• Nalidixic acid is most effective against gram-negative bacteria, including Enterobacter species and other common resistant species
• It is suitable in laboratory applications for research of the mechanisms of gyrase activity and metabolic pathways related to DNA synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-8-3374
Molecular Formula C12H11N2NaO3
Molecular Weight (g/mol) 254.22
MDL Number MFCD00064376
InChI Key ROKRAUFZFDQWLE-UHFFFAOYSA-M
Synonym nalidixic acid sodium salt, baktogram, nalidixic acid sodium, sodium nalidixate, chemiurin, dixilina, nalidixate sodium anhydrous, nalidixate sodium, nalidixate sodium anhydrous usan, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
PubChem CID 3864541
IUPAC Name sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]

Specifications

Melting Point >280°C
Color White to Yellow
Quantity 25 g
Beilstein 3580062
Solubility Information Soluble at 50mg/ml in water
Formula Weight 254.22
Physical Form Powder
Chemical Name or Material Nalidixic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.