missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic Acid, TRC

Product Code. 30007389
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007389 50 mg 50mg
30010038 100 mg 100mg
17993135 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007389 Supplier Toronto Research Chemicals Supplier No. TRCN27560050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389-08-2
Molecular Formula C12 H12 N2 O3
Molecular Weight (g/mol) 232.24
Synonym Nalidixic acid, 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid, 1,8-Naphthyridine-3-carboxylic acid, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,4-Dihydro-1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid, 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid, 1-Ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid, 1-Ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid, 3-Carboxy-1-ethyl-7-methyl-1,8-naphthyridin-4-one, Betaxina, Cybis, Dixiben, Eucistin, Innoxalomn, NSC 82174, Nacid, Nalidic acid, Nalidicron, Nalidixan, Nalidixic acid, Nalidixin, Nalidixinic acid, Nalitucsan, Nalix, Nalurin, Narigix, Naxuril, NegGram, Nelidix, Nevigramon, Nicelate, Nogram, Poleon, Specifin, Uralgin, Uriben, Uriclar, Urisal, Urodixin, Uroman, Uroneg, Uropan, Win 18320, Wintomylon
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2ccc(C)nc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H12N2O3/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14/h4-6H,3H2,1-2H3,(H,16,17)
Formula Weight 232.0848
Percent Purity >95%
Chemical Name or Material Nalidixic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.