missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Acetyl-DL-Glutamine-2,3,3,4,4-d5, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N2-Acetyl-DL-glutamine-2,3,3,4,4-D5 |
| CAS | 14341-82-3 |
| Molecular Formula | C7 D5 H7 N2 O4 |
| Quantity | 0.01 g |
| Synonym | Glutamine-2,3,3,4,4-d5, N2-acetyl-, DL- (8 CI), N2-Acetyl-DL-glutamine-2,3,3,4,4-D5, Nα-Acetyl-DL-glutamine-2,3,3,4,4-d5 |
| InChI Formula | InChI=1 S/C7H12N2O4/c1-4(10)9-5(7(12)13)2-3-6(8)11/h5H,2-3H2,1H3,(H2,8,11)(H,9,10)(H,12,13)/i2D2,3D2,5 D |
| SMILES | [2 H]C([2 H])(C(=O)N)C([2 H])([2 H])C([2 H])(NC(=O)C)C(=O)O |
| IUPAC Name | 2-acetamido-5-amino-2,3,3,4,4-pentadeuterio-5-oxopentanoic acid |
| Molecular Weight (g/mol) | 193.212 |
| Formula Weight | 193.111 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction