missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 98%, Thermo Scientific Chemicals

Product Code. 15466936
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466936 5 g 5g
15456936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466936 Supplier Thermo Scientific Alfa Aesar Supplier No. H62025.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

Solubility
Soluble in dilute acid.

Notes
Hygroscopic. Store in cool, well ventilated area. Keep container tightly closed. Store away from water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35761-26-3
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.243
MDL Number MFCD00237345
InChI Key FOXRXVSTFGNURG-VIFPVBQESA-N
Synonym z-dap-oh, cbz-beta-amino-l-alanine, 3-amino-2-n-cbz-l-alanine, s-3-amino-2-benzyloxy carbonyl amino propanoic acid, cbz-l-dap-oh, s-3-amino-2-benzyloxycarbonylamino propanoic acid, cbz-dap-oh, h-dap z-oh, n-alpha-cbz-l-2,3-diaminopropionic acid, z-dpr-oh
PubChem CID 2756313
IUPAC Name (2S)-3-amino-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CN)C(=O)O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 4486950
Solubility Information Soluble in dilute acid.
Quantity 5 g
Formula Weight 238.24
Optical Rotation −39° (c-1 in 1M HCl)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.