missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-asparagine, 95%

Product Code. 15417628
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417628 25 g 25g
15407628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417628 Supplier Thermo Scientific Alfa Aesar Supplier No. H61121.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75647-01-7 | C9H16N2O5 | 232.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75647-01-7
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00065558
InChI Key FYYSQDHBALBGHX-RXMQYKEDSA-N
Synonym boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1623168
IUPAC Name (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Beilstein 4843040
Quantity 25 g
Formula Weight 232.23
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-D-asparagine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.