missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-lysine, 98%

Product Code. 15449408
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449408 5 g 5g
15459408 250 mg 250mg
15439408 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449408 Supplier Thermo Scientific Alfa Aesar Supplier No. H62966.06
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106719-44-2 | C11H22N2O4 | 246.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106719-44-2
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00076956
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-d-lys-oh, boc-l-lys-oh, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-alpha-t-butoxycarbonyl-d-lysine
PubChem CID 7018770
IUPAC Name (2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O

Specifications

Melting Point 196°C to 200°C
Quantity 5 g
Optical Rotation −22° (c=2 in Methanol)
Formula Weight 246.3
Percent Purity 98%
Chemical Name or Material Nalpha-Boc-D-lysine

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.