missing translation for 'onlineSavingsMsg'
Learn More

NAlpha-Boc-L-histidine, TRC

Product Code. 30337231
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337231 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337231 Supplier Toronto Research Chemicals Supplier No. TRCB65622550G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nalpha-Boc-L-histidine
CAS 17791-52-5
Molecular Formula C11H17N3O4
Quantity 50 g
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-histidine, N-Carboxy-L-histidine N-tert-Butyl Ester, (+)-Nα-(tert-Butoxycarbonyl)-L-histidine, (tert-Butoxycarbonyl)-L-histidine, (tert-Butoxycarbonyl)histidine, N-(tert-Butoxycarbonyl)histidine, N-tert-Butoxycarbonyl-L-histidine, N-tert-Butyloxycarbonyl-L-histidine, NSC 334942, Nα-(tert-Butyloxycarbonyl)-L-histidine, Nα-tert-Butoxycarbonyl-L-histidine, tert-Butyloxycarbonyl-L-histidine
InChI Formula InChI=1S/C11H17N3O4/c1-11(2,3)18-10(17)14-8(9(15)16)4-7-5-12-6-13-7/h5-6,8H,4H2,1-3H3,(H,12,13)(H,14,17)(H,15,16)/t8-/m0/s1
SMILES O=C(O)[C@@H](NC(OC(C)(C)C)=O)CC1=CN=CN1
IUPAC Name (tert-butoxycarbonyl)-L-histidine
Molecular Weight (g/mol) 255.27
Formula Weight 255.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.