missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine, 95%, Thermo Scientific™

Product Code. 15499668
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15499668

Brand: Thermo Scientific Alfa Aesar H63143.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N alpha-Boc-N epsilon-trifluoroacetyl-L-lysine, is used as a fine chemical intermediate.

Solubility
Soluble in DMSO.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16965-06-3
Molecular Formula C13H21F3N2O5
Molecular Weight (g/mol) 342.32
MDL Number MFCD00037104
InChI Key DEIYNDIFGSDDCY-SVGMAFHSNA-N
Synonym boc-lys tfa-oh, s-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, boc-l-lys tfa-oh, 2s-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, ambotzbaa5790, n-a-boc-n-epsilon-trifluoroacetyl-l-lysine, n-a-boc-n-e-trifluoroacetyl-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, n∼2∼-tert-butoxycarbonyl-n∼6∼-trifluoroacetyl-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-trifluoroacetamido hexanoic acid
PubChem CID 7009540
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O

Specifications

Color White
Solubility Information Soluble in DMSO.
Quantity 25 g
Formula Weight 342.31
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.