missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine, 98%

Product Code. 15428618
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428618 25 g 25g
15438618 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428618 Supplier Thermo Scientific Alfa Aesar Supplier No. H62019.14
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 2483-46-7 | C16H30N2O6 | 346.42 g/mol

Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

Solubility
Soluble in chloroform, ether, ethyl acetate and methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2483-46-7
Molecular Formula C16H30N2O6
Molecular Weight (g/mol) 346.42
MDL Number MFCD00038515
InChI Key FBVSXKMMQOZUNU-LDGXTIHJNA-N
Synonym boc-lys boc-oh, s-2,6-bis-tert-butoxycarbonylaminohexanoic acid, boc-lys boc, s-2,6-bis tert-butoxycarbonyl amino hexanoic acid, di-boc-l-lysine, na, ne-bis-boc-l-lysine, boc-l-lys boc-oh, boc-l-lys boc, boc-l-lysine boc, s-2,6-bis-tert-butoxycarbonylamino-hexanoic acid
PubChem CID 7349648
IUPAC Name (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Color White
Solubility Information Soluble in chloroform,ether,ethyl acetate and methanol
Quantity 25 g
Formula Weight 346.42
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.