missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NAMPT PROTAC™ A7
GREENER_CHOICE

Product Code. 30000028
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000028 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000028 Supplier Tocris Bioscience™ Supplier No. 7842/5
  • ONLINE EXCLUSIVE PRICE
    429.00 €
    405.30 € / 5mg
    Save 23.70 €
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective nicotinamide phosphoribosyl transferase (NAMPT) Degrader (PROTAC™;)

NAMPT PROTAC A7 is a potent and selective intracellular nicotinamide phosphoribosyl transferase (iNAMPT) Degrader (PROTAC™;) (IC50 of 9.5 nM against the enzymatic activity of NAMPT). Comprises a potent NAMPT inhibitor joined by a linker to a ligand for Von Hippel-Lindau (VHL) protein. Proteomic analysis of global protein level changes confirms selectivity. NAMPT PROTAC A7 decreases iNAMPT level and the secretion of extracellular NAMPT (eNAMPT) in CT26 or MC38 cells. In CT26 tumor bearing BALB/c mice, the compound degrades tumoral NAMPT, and boosts antitumor immunity by inhibiting tumor infiltrating myeloid-derived suppressive cells.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4R)-1-((S)-18-(tert-Butyl)-1,16-dioxo-1-(4-((4-((4-(3-(pyridin-3-ylmethyl)thioureido)phenyl)sulfonyl)piperazin-1-yl)methyl)phenyl)-5,8,11,14-tetraoxa-2,17-diazanonadecan-19-oyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-ca
CAS 2484876-94-8
Quantity 5 mg
Target Active Degraders
Molecular Formula C58H76N10O11S3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.