missing translation for 'onlineSavingsMsg'
Learn More

Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 647, Chromotek

Product Code. 18058695
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18058695 100 μL 100µL
18006865 10 μL 10µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18058695 Supplier Chromotek Supplier No. shuGCL6472100
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alpaca Recombinant Monoclonal Antibody

Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.

Specifications

Antigen Nano-Secondary™
Conjugate CoraLite Plus 647
Host Species Alpaca
Quantity 100 μL
Primary or Secondary Secondary
Target Species Human
Form Liquid
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.