missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nano Silicon Glass Plates

Product Code. 10206761 Shop All Macherey-Nagel Products
Change view
Click to view available options
Description:
Degree of Silanisation: 100%
Degree of Silanisation: 50%
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10206761 Degree of Silanisation: 100% Pack of 25
10545381 Degree of Silanisation: 50% Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10206761 Supplier Macherey-Nagel™ Supplier No. 811062
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

HPTLC glass plates in different plate sizes for many different applications. Silica gel with octadecyl (C18, ODS, RP18) modification. Carbon content: approx. 14 %.

HPTLC Glass Plates modified silica gel, RP, Nano-SIL C18-100 UV254

  • Base material: glass plates with silica gel
  • Mode: Reversed Phase
  • Layer Hardness: Standard, Soft

Adsorbent:

  • Silica Gel with octadecyl (C18, ODS, RP18) modification
  • Phase Nano-SIL C18-100 UV254
  • UV Indicator: Yes
  • Particle Type: Fully porous particles (FPP)
  • Particle Shape: Irregular
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 2-10 μm
  • Binder: Yes

Indicators:

This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).

Binder:

Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Recommended Applications:

Alkaloids, Amino acids, Barbiturates, Drugs, Flavonoids, Indole derivatives, Optical brighteners, Peptides, Phenols, Polycyclic aromatic hydrocarbons (PAH), Preservatives, Reversed phase (RP), Steroids, Vegetable oils (identification of fatty oils acc. to EP)

General TLC Glass Plate Features from Macherey-Nagel:

  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
  • The plate can be easily labelled for clear identification
  • Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
  • Increased separation efficiency due to an optimized particle size distribution
  • A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber

Specifications

Backing Material Nano Silica
Description Degree of Silanisation: 100%
Material Nano Silicon
Particle Size 2 to 10 μm
Pore Size 60 Å
Quantity 25
Type Glass Plate
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface 500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 10 x 10 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.