missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The use of rectangular cuvettes with 50 mm optical path allows the detection of least amounts of the detectable substance.
- The use of a large sample volume guarantees safe and reproducible results.
- Approved - reliable and comparable results.
- The reaction basis is based on internationally approved standard methods like DIN-, EN-, ISO-, EPA- und APHA.
- Versatile - the perfect test for every request.
- For all important parameters in water and wastewater analysis.
- Precise - easy handling and highest accuracy.
- All tests contain accurate, ready-to-use, predosed reagents.
- Unique - no mix up of tests.
- Every test box is equipped with a color-coded label and can be clearly identified by its color.
Specifications
Specifications
| Analytes | Hydrazine, L N2H4 |
| Description | Seawater suitable |
| No. of Tests | 250 Determinations |
| Range | 0.002 to 1.50 mg/L |
| Test Method | Standard Test 1-44. Photometric determination with 4-dimethylaminobenzaldehyde |
| Type | Photometric water analysis test |
Safety and Handling
Product Identifier
- NANOCOLOR Hydrazine
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contain: sulfuric acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?