Learn More
Thermo Scientific™ NanoDrop Label Printer - DYMO LabelWriter 550
Product Code. 31921060 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
DYMO LabelWriter 550 Turbo w/Labels
New sales of the NanoDrop Label Printer - DYMO LabelWriter 450 have been discontinued. Please see the newest model: NanoDrop Label Printer - DYMO LabelWriter 550 Turbo.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
31921060 New sales of the NanoDrop Label Printer - DYMO LabelWriter 450 have been discontinued. Please see the newest model: NanoDrop Label Printer - DYMO LabelWriter 550 Turbo. Each
31921047 DYMO LabelWriter 550 Turbo w/Labels Each
31034392 DYMO LabelWriter 550 Turbo w/Labels -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31921060 Supplier Thermo Scientific™ Supplier No. PNTRLW400
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Thermo Scientific™ NanoDrop™ Dymo LabelWriter 450 Printer is a convenient accessory for NanoDrop instruments. It prints sample data onto self-adhesive labels, which are included with purchase of printer.
The Dymo LabelWriter 550 Turbo label printer with labels for the Thermo Scientific™ NanoDrop™ is a convenient accessory for our microvolume UV-Vis instruments. It quickly and easily prints sample data onto self-adhesive labels, which are included with purchase of printer.
Features include:
  • Generates 2.31 x 4 inch (59mm x 102mm) self-adhesive labels, allowing you to place labels directly in laboratory notebooks
  • Prints results for single samples or batches of samples
  • Direct thermal label printer, no ink or toner required
LabelWriter is a registered trademark of DYMO Corporation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description New sales of the NanoDrop Label Printer - DYMO LabelWriter 450 have been discontinued. Please see the newest model: NanoDrop Label Printer - DYMO LabelWriter 550 Turbo.