Learn More
Thermo Scientific™ NanoDrop™ Eight Pipette Guide
Product Code. 31921069 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31921069 NanoDrop Eight Spectrophotometers Each
1 options
Product Code. 31921069 Supplier Thermo Scientific™ Supplier No. 840343600
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Thermo Scientific™ NanoDrop™ Eight Pipette guide allows you to comfortably and accurately pipette samples onto all eight wells of the instrument using an 8-channel precision pipettor.
The Thermo Scientific™ NanoDrop™ Eight Pipette guide allows you to comfortably and accurately pipette samples onto all eight wells of the instrument using an 8-channel precision pipettor.
The pipette guide is magnetic and movable from side-to-side allowing left- and right-handed users to quickly and comfortably dispense samples
Specifications
Description NanoDrop Eight Pipette Guide
For Use With (Equipment) NanoDrop Eight Spectrophotometers