missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop Lite Plus Spectrophotometers

Product Code. 31921085 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
ND Lite Plus
Printer Included:
No
Yes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Printer Included Unit Size
31921085 ND Lite Plus No Each
31921094 ND Lite Plus Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31921085 Supplier Thermo Scientific™ Supplier No. NDLPLUSGL
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quickly and easily assess sample concentration and purity ratios with the entry-level Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer. The instrument delivers accurate measurements in an easy-to-use, affordable package. Ideal for teaching and busy research labs.

Quickly and easily assess sample concentration and purity ratios with the Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer, the entry-level complement to the full-featured Thermo Scientific™ NanoDrop™ One/OneC and NanoDrop™ Eight Spectrophotometers. While offering fewer features than our premium models, this instrument delivers fast, accurate measurements in an easy-to-use, affordable package.

The NanoDrop Lite Plus Spectrophotometer is a valuable teaching tool and is ideal for quality check measurements in busy research labs as well. Measure purified DNA, RNA, and protein concentration up to 30 Abs and calculate critical A260/A280 and A260/A230 purity ratios using trusted NanoDrop technology.

Spectrophotometer for labs
The NanoDrop Lite Plus Spectrophotometer is designed for routine measurements of nucleic acid and protein samples using only 1–2 μL. Our pioneering sample-retention technology eliminates the need to dilute highly concentrated samples, making measurements easy. Simply pipette your sample directly onto the optical measurement pedestal, measure, and wipe clean with a lint-free wipe. Its compact design, with built-in controls and software, means the NanoDrop Lite Plus Spectrophotometer takes up little space on your benchtop.

With an intuitive user interface and modern industrial design, the NanoDrop Lite Plus Spectrophotometer is the instrument of choice for basic microvolume measurements. Even if budgets are tight, your lab can have a NanoDrop, too.

Confidence in your results

  • Standard measurements
    • Measures purified DNA, RNA, and protein concentration up to 30 Abs
      • Up to 1,500 ng/μL dsDNA
      • Up to 1,200 ng/μL RNA
      • Up to 45 mg/mL BS
    • Calculates critical A260/A280 and A260/A230 purity ratios
  • Fast, accurate measurements from only 1–2 μL—saves precious samples, no waste
  • Sample-retention technology uses surface tension to hold the sample in place—no cuvettes or expensive consumables are needed
  • Intuitive interface
    • Built-in touchscreen operation
    • No computer required
  • Data storage and transfer
    • Last 1,000 measurements saved locally; export data to a USB device
  • Optional printer available for freezer-compatible labels (not included)
  • Compact and portable—fits on any lab bench, ready for your next experiment
  • Affordably priced—stretch tight budgets

Reach your goals

  • Academic teaching—educate students on molecular biology principles and experimental techniques on an industry-leading instrument
  • Academic research labs—facilitate your research and publication of your novel research findings
  • Biotech start-ups—accelerate the develop of new products

Specifications

Absorbance Accuracy 3% at 0.97 A, 302 nm, 23 ± 2°C Abs/mm at 25 °C
Absorbance Range 0.04 to 30 Abs (10 mm equivalent)
Certifications/Compliance UL/CSA and CE
Compatibility Local Control Only
Concentration 1,500 ng/μL (1,200 ng/μL) dsDNA (RNA)45 mg/mL (21 mg/mL) BSA (lgG)
Connections USB 2.0 Port
Depth (Metric) 22 cm
Detection Limits 2.0 ng/μL (1.6 ng/μL) dsDNA (RNA)0.06 mg/mL (0.03 mg/mL) BSA (lgG)
Detector Type 2048-element CMOS Linear Image Sensor
Display LCD Touchscreen
Light Source Xenon Flash
Product Line Microvolume
Type Microvolume UV-Vis Spectrophotometer
Voltage 100/240 V
Wavelength Range 230 nm, 260 nm, 280 nm
Footprint 27 x 22 x 22 cm (0.6 x 8.7 x 8.7 in); W x D X H
For Use With (Application) Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Height (Metric) 22 cm
Includes ND Lite Plus
Material Stainless Steel (303), Quartz Fiber
Measurement Time ≤ 5 seconds
No. of Samples 1
Pathlength 1.0 mm and 0.2 mm (auto adjust)
Power Consumption 18 W (standby < 3 Watts)
Printer Included No
Processor Standalone control with data export via USB Flash Drive
Sample Volume (Metric) 1 to 2 μL
Spectral Resolution ≤ 1.8 nm (FWHM at Hg 254 nm)
System Requirements Standalone control with data export via USB Flash Drive
Warranty Two year
Wavelength Accuracy ± 1 nm
Weight (English) 6.0 lbs (without printer)
Weight (Metric) 2.7 kg (without printer)
Width (Metric) 27 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.