Learn More
Implen™ NanoPhotometer™ 21 CFR Part 11 Software Packages
Product Code. 16308747 Shop All Implen Products
Change view
Click to view available options
Includes:
Software License Key
Unit Size:
Each
Product Code. Includes unitSize
16308747 Software License Key Each
1 options
Product Code. 16308747
Brand: Implen™ CFR21SW
CFR21 Software complies with FDA 21 CFR Part 11 requirements, and is an optional software tool that is ideal for GxP laboratories which require proper electronic record keeping.
  • License key provided on USB flash drive
  • Software package includes user management, access control, electronic signatures, data integrity, security, and audit trail functionality
Specifications
Certifications/Compliance FDA 21 CFR Part 11
Description Software Upgrade for NanoPhotometer
For Use With (Equipment) NanoPhotometer™ N120, NP80, N60, and C40
Includes Software License Key
System Requirements Windows or Mac, Touchscreen Compatible