missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ C40 Cuvette UV-VIS Spectrophotometer

Product Code. 15598783 Shop All Implen Products
Change view
Click to view available options
Type:
Spectrophotometer
Spectrophotometer Int Battery
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15598783 Spectrophotometer Each
15528783 Spectrophotometer Int Battery Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15598783 Supplier Implen™ Supplier No. C40Touch
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cuvette applications. Suitable for all common photometric measurements like protein assays, kinetics and standard curve. Intuitive touchscreen operation, small footprint and network integration for convenient lab bench operation. Recalibration-free patented technology.

The Implen NanoPhotometer™ C40 UV/VIS spectrophotometer offers full scan data from 200 - 900 nm for all cuvette based photometric applications in Life Sciences. It can be upgraded to NanoVolume using the Implen Submicoliter Cell. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.

No recalibration is necessary over the entire lifetime. Optional CFR21 software available for 21 CFR part 11 compliance for GxP environment.

  • Detection range dsDNA 0.1 ng/μL - 130 ng/μL
  • Detection range BSA 0.003 mg/mL - 3.7 mg/mL
  • Photometric Range 0 - 2.6 A
  • Scan range 200 - 900 nm
  • Center Height (Z-Height) 8.5 mm
  • Suitable for all cuvettes with outside dimensions 12.5 x 12.5 mm
  • Temperature controlled cuvette port 37°C
  • Measure time for full scan range: 2.5 seconds
  • Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA, Lowry), kinetics, OD600 and manual methods
  • Recalibration-free due to True Path Technology™
  • Sample ControlTM and Blank ControlTM for analysis of sample quality and identification of contaminants
  • Control options: Onboard with built-in 7” Touchscreen, Computer and Tablet
  • In and Output Ports 2x USB A, USB B, HDMI, Ethernet, Wi-Fi
  • 21 CFR part 11 compliant software (optional)
  • Built-in file server
  • LIMS integration through REST API
  • Built-in battery pack, operation time: up to 8 h
  • Made in Germany

Specifications

Absorbance Accuracy < 1.75 % at 0.7 A (0.67 mm path) at 280 nm of the reading
Absorbance Range 0 to 2.6 A
Beam Center Height 8.5 mm
Connections 2x USB A, USB B, HDMI, Ethernet, WiFi
Detector Type 3648-element CCD Array
Display 7 in. Glove Compatible Touchscreen, 1024 x 600 pixels
Dimensions (L x W x H) 200 x 200 x 120 mm
Languages English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
Light Source Xenon Flash
Spectral Bandwidth <1.8 nm
Type Spectrophotometer
Voltage 90/250 V
Wavelength Range 200 to 900 nm
For Use With (Application) Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom
Includes Power Supply, USB Cable, Dust Cover, Software and Manual
Measurement Time 2.5 sec
No. of Samples 1
Operating Temperature 37 °C ± 0.5 °C
Pathlength 10, 5, 2, 1, 0.5 mm
Processor Linux based NPOS
System Requirements Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS
Wavelength Accuracy ± 0.75 nm
Weight (Metric) 4.2 kg
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.