missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ NP80 Nano-Volume and Cuvette UV-VIS Spectrophotometer

Product Code. 15485824 Shop All Implen Products
Change view
Click to view available options
Model:
NP80-Mobile
NP80-Touch
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15485824 NP80-Touch Each
15512465 NP80-Mobile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485824 Supplier Implen™ Supplier No. NP80Touch

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

The Implen NanoPhotometer™ NP80 UV/Vis spectrophotometer for NanoVolume and cuvette applications. Quantify DNA, RNA, proteins and scan other samples within seconds.

The flexible Implen NanoPhotometer™ NP80 UV/VIS spectrophotometer offers both cuvette and NanoVolume measurements. It provides full scan data from 200 - 900 nm for various Life Science applications like quantification of nucleic acids, proteins or cell cultures. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.

Disturbing surface tension and evaporation effects are eliminated due to the compression of the sample. No recalibration is necessary over the entire lifetime. Optional CFR21 software available for 21 CFR part 11 compliance for GxP environment.

  • Detection range dsDNA 1 ng/μL - 16,500 ng/μL
  • Detection range BSA 0.03 mg/mL - 478 mg/mL
  • Minimum sample Size 0.3 μL
  • Scan range 200 - 900 nm
  • Cuvette Performance 0 - 2.6 A
  • Integrated Vortex for tubes up to 2.0 mL
  • Measure time for full scan range: 2.5 - 4.0 seconds
  • Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA, Lowry), kinetics, OD600 and manual methods
  • Auto Sample function starts automatically once sample arm is closed
  • Recalibration-free due to True Path Technology™
  • Independent from surface tension of sample due to Sample Compression Technology™
  • Sample Control™ and Blank Control™ for analysis of sample quality and identification of contaminants
  • Control options: Onboard with built-in 7 in. Touchscreen, Computer and Tablet
  • In & Output Ports 2x USB A, USB B, Ethernet, Wi-Fi
  • 21 CFR part 11 compliant software (optional)
  • Built-in file server
  • LIMS integration through REST API
  • Made in Germany

Specifications

Absorbance Accuracy < 1.75% at 0.7 A at 280 nm of the reading
Absorbance Range 0.02 to 330 A NanoVolume, 0 to 2.6 A Cuvette
Beam Center Height 8.5 mm
Connections 2x USB A, USB B, Ethernet, WiFi
Detector Type 3648-element CCD Array
Display 7 in. Glove Compatible Touchscreen, 1024 x 600 pixels
Dimensions (L x W x H) 20 x 20 x 12 cm
Languages English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
Light Source Xenon Flash
Spectral Bandwidth <1.8 nm
Voltage 90/250 V
Wavelength Range 200 to 900 nm
For Use With (Application) Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom
Includes Power Supply, USB Cable, Dust Cover, Software & Manual
Measurement Range 1 to 16,500 ng/μL
Measurement Time 2.5 to 4.0 sec
Model NP80-Touch
No. of Samples 1
Operating Temperature 37 °C ± 0.5 °C Cuvette
Pathlength 0.67 and 0.07 mm
Processor Linux based NPOS
Sample Volume (Metric) 0.3 to 2 μL
Stray Light < 0.5% @ 240 nm using NaI
System Requirements Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS
Wavelength Accuracy ± 0.75 nm
Weight (Metric) 5 kg
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.