Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Product Code. 17134791
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
300 kDa
Includes:
100/Pk.
24/Pk.
500/Pk.
Color:
Blue
Clear
Gray
Orange
Red
Unit Size:
Pack of 100
Pack of 24
Pack of 500
Product Code. 17134791
Brand: Cytiva OD100C35
Only null left
Add to basket
Add to basket
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Description EFA: 0.3 cm2
Material (Membrane) PES
MWCO 100 kDa
Sterility Non-sterile
Housing Material Polypropylene
Includes 500/Pk.
Volume (Metric) 50 to 500 μL (Sample)