missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Nanosep™ Centrifugal Devices with wwPTFE membrane

Product Code. 17174801
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Includes:
100/Pk.
500/Pk.
Unit Size:
Pack of 100
Pack of 500
Product Code. Pore Size Includes unitSize
17174801 0.2 μm 100/Pk. Pack of 100
17184801 0.2 μm 500/Pk. Pack of 500
17194801 0.45 μm 100/Pk. Pack of 100
17104811 0.45 μm 500/Pk. Pack of 500
4 options
This item is not returnable. View return policy

Product Code. 17174801

Brand: Cytiva ODPTFE02C34

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.

  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description EFA: 0.3 cm2
Material (Membrane) PTFE (Hydrophilic)
Pore Size 0.2 μm
Sterility Non-sterile
Housing Material Polypropylene
Includes 100/Pk.
Volume (Metric) 50 to 500 μL (Sample)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.