missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Nanosep™ MF Centrifugal Devices with Bio-Inert™ Membrane

Product Code. 17114801
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Includes:
100/Pk.
24/Pk.
500/Pk.
Unit Size:
Pack of 100
Pack of 24
Pack of 500
Product Code. Pore Size Includes unitSize
17114801 0.2 μm 500/Pk. Pack of 500
17124801 0.45 μm 24/Pk. Pack of 24
17104801 0.2 μm 100/Pk. Pack of 100
17194791 0.2 μm 24/Pk. Pack of 24
17134801 0.45 μm 100/Pk. Pack of 100
17144801 0.45 μm 500/Pk. Pack of 500
6 options
This item is not returnable. View return policy

Product Code. 17114801

Brand: Cytiva ODM02C35

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.

  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Color Aqua
Description EFA: 0.3 cm2
Material (Membrane) Bio-Inert (modified nylon)
Pore Size 0.2 μm
Sterility Non-sterile
Housing Material Polypropylene
Includes 500/Pk.
Volume (Metric) 50 to 500 μL (Sample)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.