missing translation for 'onlineSavingsMsg'
Learn More

Biochrom NanoVue Plus Spectrophotometer

Product Code. 10506405
Change view
Click to view available options
Pathlength:
0.2 mm or 0.5 mm (automatic or manual)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pathlength unitSize
10506405 0.2 mm or 0.5 mm (automatic or manual) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10506405 Supplier Biochrom Supplier No. 28956057

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Easy to use instrument designed for reliable measurements of nucleic acids and proteins. Operates without the use of a PC.

  • IQ/OQ Documents available
  • Gold-colored sample plate enhances compatibility with the wide range of chemicals and sample types used in today's busy laboratories.
  • Predefined methods for protein quantitation, including direct UV, Bradford, BCA, Biuret, and Lowry, with capability to run up to 27 standards.
  • Accurately assays low volumes of 0.5 to 5μL conserving precious samples.
  • Two pathlengths - 0.2mm and 0.5mm
  • Convenient, drop and read sample plate eliminates the need for cuvettes and capillaries.
  • Predefined methods for nucleic acid quantitation, including concentration and purity.
  • UV-Visible spectrophotometer for quick and accurate quantitation of nucleic acids and protein samples.
  • No PC: Large, high-resolution graphical display enables a quick read of relevant results.
  • Calibration curves, kinetics or ratio measurements are all displayed at the touch of a button.
  • Hydrophobic plate reduces sample carry over and allows for excellent sample recovery.

Specifications

Absorbance Range Max. ±1% at 259 nm (at 0.7 to 0.8 A using uracil)
Detector Type Twin CCD Array
Dimensions (L x W x H) 260 x 390 x 100 mm
Light Source Xenon
Spectral Bandwidth 5 nm
Type Spectrophotometer
Wavelength Range 200 to 1100 nm
Electrical Requirements 100/240 V ± 10%, 50/60 Hz, 50 V
Pathlength 0.2 mm or 0.5 mm (automatic or manual)
Wavelength Accuracy ±2 nm
Wavelength Repeatability ±0.5 nm
Weight (Metric) 4.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.