Learn More
Tocris Bioscience™ Napabucasin
STAT3 inhibitor; also blocks cancer stem cell self-renewal
Brand:  Tocris Bioscience™ 5522/50
Encompass
Product Code. 15766896
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Napabucasin is a STAT3 inhibitor. Blocks spherogenesis of cancer stem cells (CSC), but not hematopoietic stem cells. Inhibits PaCa-2 xenograft tumor growth in mice, even after cessation of treatment, and decreases population of CSCs in PaCa-2 tumors from treated mice. Also blocks spleen and liver metastases of colon cancer cells in a mouse model.
Specifications
Specifications
2-Acetylnaphtho[2,3-b]furan-4,9-dione
83280-65-3