missing translation for 'onlineSavingsMsg'
Learn More

Naphazoline Hydrochloride, TRC

Product Code. 30473843
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473843 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473843 Supplier Toronto Research Chemicals Supplier No. TRCN3439505G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naphazoline Hydrochloride
CAS 550-99-2
Molecular Formula C14 H14 N2 . Cl H
Quantity 5 g
Synonym Naphazoline Hydrochloride, 4,5-Dihydro-2-(1-naphthalenylmethyl)-1H-imidazole monohydrochloride, 2-(1-Naphthylmethyl)-2-imidazoline monohydrochloride, Naphazoline hydrochloride, 2-(α-Naphthylmethyl)-2-imidazoline hydrochloride, 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 2-(1-Naphthylmethyl)imidazoline hydrochloride, Ak-Con, Albalon, Allerest Eye Drops, Clera, Coldan, Comfort Eye Drops, Degest 2, Iridina Due, Naphazoline chloride, Naphcon, Naphthasolium chloride, Niazol, Ocumethyl, Opcon, Privine (American), Privine hydrochloride, Prizole hydrochloride, Rhinantin, Rhinatin, Rhinoperd, Sanorin-Spofa, Strictylon, Stricylon, Vasoclear, Vasocon
InChI Formula InChI=1S/C14H14N2.ClH/c1-2-7-13-11(4-1)5-3-6-12(13)10-14-15-8-9-16-14;/h1-7H,8-10H2,(H,15,16);1H
SMILES Cl.C(C1=NCCN1)c2cccc3ccccc23
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
Molecular Weight (g/mol) 246.74
Formula Weight 246.0924
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.