missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical

Product Code. 10294100 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10294100 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10294100 Supplier Fisher Chemical Supplier No. N/0216/46
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 532-02-5
Color White
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C10H7NaO3S
MDL Number MFCD00064186
Synonym sodium 2-naphthalenesulfonate, sodium naphthalene-2-sulfonate, 2-naphthalenesulfonic acid sodium salt, 2-naphthalenesulfonic acid, sodium salt, unii-r5f0ctd2oj, sodium-2-naphthalenesulfonate, sodium naphthalene-6-sulfonate, 2-naphthalene sulfonic acid sodium salt, sodium beta-naphthalenesulfonate, sodium naphthalene-2-sulphonate
InChI Key YWPOLRBWRRKLMW-UHFFFAOYSA-M
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1
IUPAC Name sodium;naphthalene-2-sulfonate
Molecular Weight (g/mol) 230.21
PubChem CID 23661868
Formula Weight 230.22g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • 2-Naphthalenesulfonic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.