missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene Black 10B, Technical, Fisher Chemical

Product Code. 10528440 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass Jar
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10528440 10 g Glass Jar 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528440 Supplier Fisher Chemical Supplier No. N/0211/45

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1064-48-8
Color Brown
pH 9
Packaging Glass Jar
Molecular Formula C22H14N6Na2O9S2
MDL Number 4017
Quantity 10 g
Synonym amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, parent, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 616.487
PubChem CID 44134531
Formula Weight 616.48g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.