missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-d8, for NMR, 98+ atom % D

Product Code. 10647504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647504 5 g Glass Bottle 5g
10605052 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10647504 Supplier Thermo Scientific Acros Supplier No. 174960050
  • ONLINE EXCLUSIVE PRICE
    294.00 €
    260.00 € / 5g
    Save 34.00 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1146-65-2 | C10H8 | 136.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1146-65-2
Molecular Formula C10H8
Molecular Weight (g/mol) 136.22
MDL Number MFCD00001743
InChI Key UFWIBTONFRDIAS-PGRXLJNUSA-N
Synonym naphthalene-d8, 2h8 naphthalene, perdeuteronaphthalene, naphthalene d8, perdeuterated naphthalene, 2 h? naphthalene, naphthalene-d8, analytical standard, naphthalene-d8 >98.0 atom % d, naphthalene d8 10 microg/ml in cyclohexane
PubChem CID 92148
IUPAC Name 1,2,3,4,5,6,7,8-octadeuterionaphthalene
SMILES [2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H]

Specifications

Physical Form Crystals or Crystalline Powder
Color White
Deuterium (D) 98.0% min.
Flash Point 78°C
Packaging Glass Bottle
Quantity 5 g
Melting Point 81.0°C to 83.0°C
Linear Formula C10D8
Formula Weight 136.22
Grade NMR
Chemical Name or Material Naphthalene-d8, For NMR
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Naphthalene-d8
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Carcinogenicity Category 2
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.