missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-d8, TRC

Product Code. 30472212
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30472212 Supplier Toronto Research Chemicals Supplier No. TRCN3456041G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naphthalene D8
CAS 1146-65-2
Purity Grade Notes HPLC
Molecular Formula C10 D8
Quantity 1 g
Synonym Naphthalene-1,2,3,4,5,6,7,8-d8, Naphthalene-d8 (6CI,8CI,9CI), Naphthalene (C10D8), Octadeuterionaphthalene, Octadeuteronaphthalene, Perdeuterated naphthalene, Perdeuterionaphthalene, Perdeuteronaphthalene, Naphthalene D8
InChI Formula InChI=1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D
SMILES [2H]c1c([2H])c([2H])c2c([2H])c([2H])c([2H])c([2H])c2c1[2H]
IUPAC Name 1,2,3,4,5,6,7,8-octadeuterionaphthalene
Molecular Weight (g/mol) 136.22
Formula Weight 136.1128
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.