missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, TRC

Product Code. 18013820
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18013820 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18013820 Supplier Toronto Research Chemicals Supplier No. TRCN34560050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 91-20-3
Molecular Formula C10 H8
Quantity 50 g
Synonym Naphthalene, Albocarbon, Dezodorator, Moth flakes, NSC 37565, Naphthene, Tar camphor, White tar
InChI Formula InChI=1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H
SMILES c1ccc2ccccc2c1
IUPAC Name naphthalene
Molecular Weight (g/mol) 128.17
Formula Weight 128.0626
Chemical Name or Material Naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.