missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Naphthol Green B, pure

Product Code. 10774081 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10774081

Brand: Thermo Scientific Chemicals 152931000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19381-50-1
Molecular Formula C30H18FeN3Na3O15S3
Molecular Weight (g/mol) 881.474
MDL Number MFCD00003886
InChI Key VLBLPLXOYXEXJK-UHFFFAOYSA-K
Synonym Acid Green 1, C.I. 10020
PubChem CID 14598749
IUPAC Name trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron
SMILES C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]

Specifications

Beilstein 11, 332
Solubility Information Solubility in water: 30 mg/mL water. Other solubilities: 0.9 mg/mL ethanol, 9 mg/mL ethylene glycol monomethyl ether
Infrared Spectrum Authentic
Formula Weight 878.47
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Green
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Naphthol Green B
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.