missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Naphthol Green B

Product Code. 11443817
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11443817

Brand: Thermo Scientific Chemicals A18268.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol

It finds its applications as a leather and fur dye and as a biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its applications as a leather and fur dye and as a biological stain.

Solubility
Soluble in water (160 mg/ml at 20°C), ethanol (0.9 mg/ml), and ethylene (9 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19381-50-1
Molecular Formula C30H18FeN3Na3O15S3
Molecular Weight (g/mol) 881.474
MDL Number MFCD00003886
InChI Key VLBLPLXOYXEXJK-UHFFFAOYSA-K
Synonym Acid Green 1; C.I. 10020
PubChem CID 14598749
IUPAC Name trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron
SMILES C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]

Specifications

Solubility Information Soluble in water (160mg/ml at 20°C),ethanol (0.9mg/ml),and ethylene (9mg/ml).
Formula Weight 878.47
Quantity 25 g
Chemical Name or Material Naphthol Green B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.